missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-DHMEQ, MedChemExpress
Description
(-)-DHMEQ (Dehydroxymethylepoxyquinomicin) is a potent, selective and irreversible NF-κB inhibitor that covalently binds to a cysteine residue. (-)-DHMEQ inhibits nuclear translocation of NF-κB and shows anti-inflammatory and anticancer activity.
Specifications
Specifications
| Chemischer Name oder Material | (-)-DHMEQ |
| CAS | 287194-40-5 |
| Farbe | Gray |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C13H11NO5 |
| Menge | 10 mg |
| Synonym | Dehydroxymethylepoxyquinomicin |
| Löslichkeitsinformationen | DMSO : 50 mg/mL (191.40 mM; Need ultrasonic) |
| SMILES | O=C1[C@@H](O2)[C@@H]2[C@@H](O)C(NC(C3=C(O)C=CC=C3)=O)=C1 |
| Molekulargewicht (g/mol) | 261.23 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?