Learn More
STEMCELL Technologies DisCoZyme 2, ACF
Artikelnummer. 17149271
Encompass_Preferred
Click to view available options
Menge:
50 mg
Packungsgröße:
50 Milligramm
Artikelnummer. 17149271
Marke: STEMCELL Technologies 07450
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Collagenase/dispase blend with high collagenase and high caseinase activity
  • DisCoZyme 2, Animal Component-Free (ACF) is a blend of collagenase and dispase, both of which are obtained from cultures free of animal-derived materials
  • DisCoZyme 2, ACF contains a minimum of 250 collagenase units/mg and 2000 caseinase units/mg of dry weight
  • Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
  • Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
  • Dispase is a neutral, amino-endoprotease that cleaves the N-terminal peptide bond of non-polar amino acid residues
  • Its proteolytic activity also allows it to maintain membrane integrity
  • DisCoZyme 2, ACF is designed to contain high levels of collagenase and caseinase activity and is intended for use in applications where prevention of the introduction of potential animal-derived pathogens is necessary
Spezifikation
Menge 50 mg
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.