Learn More
STEMCELL Technologies DisCoZyme 2, ACF
Click to view available options
Menge:
50 mg
Packungsgröße:
50 Milligramm
Beschreibung
- DisCoZyme 2, Animal Component-Free (ACF) is a blend of collagenase and dispase, both of which are obtained from cultures free of animal-derived materials
- DisCoZyme 2, ACF contains a minimum of 250 collagenase units/mg and 2000 caseinase units/mg of dry weight
- Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
- Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
- Dispase is a neutral, amino-endoprotease that cleaves the N-terminal peptide bond of non-polar amino acid residues
- Its proteolytic activity also allows it to maintain membrane integrity
- DisCoZyme 2, ACF is designed to contain high levels of collagenase and caseinase activity and is intended for use in applications where prevention of the introduction of potential animal-derived pathogens is necessary
Spezifikation
Spezifikation
| Menge | 50 mg |