missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)triphenylphosphoniumchlorid, 97 %, Thermo Scientific™

Catalog Number p-7024578
Click to view available options
Menge:
5 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
17577-28-5
C22H22ClO2P
384.84
MFCD00011746
DJGHVEPNEJKZBF-UHFFFAOYSA-M
carboethoxymethyl triphenylphosphonium chloride hydrate, 2-ethoxy-2-oxoethyl triphenylphosphonium chloride hydrate, acmc-20apm6, carbethoxymethyl-triphenylphosphonium chloride, 2-ethoxy-2-oxo-ethyl-triphenyl-phosphonium chloride, 2-ethoxy-2-oxoethyl triphenylphosphanium hydrate chloride
71317493
(2-ethoxy-2-oxoethyl)-triphenylphosphanie;chlorid;hydrat
[Cl-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
17577-28-5
C22H22ClO2P
384.84
MFCD00011746
DJGHVEPNEJKZBF-UHFFFAOYSA-M
carboethoxymethyl triphenylphosphonium chloride hydrate, 2-ethoxy-2-oxoethyl triphenylphosphonium chloride hydrate, acmc-20apm6, carbethoxymethyl-triphenylphosphonium chloride, 2-ethoxy-2-oxo-ethyl-triphenyl-phosphonium chloride, 2-ethoxy-2-oxoethyl triphenylphosphanium hydrate chloride
71317493
(2-ethoxy-2-oxoethyl)-triphenylphosphanie;chlorid;hydrat
[Cl-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (Ethoxycarbonylmethyl)triphenylphosphonium chloride
Schmelzpunkt ∼145°C (decomposition)
Menge 5 g
Beilstein 3924139
Empfindlichkeit Hygroscopic
Formelmasse 384.84
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'