missing translation for 'onlineSavingsMsg'
Learn More

FAM3C Mouse anti-Human, Clone: 2E10D5, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 60282-1-IG-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887345

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 21-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.
Spezifikation

Spezifikation

FAM3C
Monoclonal
0.62 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92520
FAM3C
FAM3C Fusion Protein Ag5516
20 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
2E10D5
Unconjugated
FAM3C
FAM3C, GS3786, ILEI, Interleukin like EMT inducer, Protein FAM3C
Mouse
Protein G
RUO
10447
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote