Learn More
Restek™ Fingertight Column Nuts
Allows wrench-free column installations.
Marke: Restek™ 21040
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Made from high-quality stainless steel
- Available for standard 0.0625 in. and compact (Agilent-style) ferrules
Spezifikation
Mutter | |
Compact Agilent-Style Ferrules for HP GCs |