Learn More
Fisherbrand™ Sharps-A-Gator™ Sharps Containers
Puncture-resistant with wide rectangular opening for horizontal loading of needles
Marke: Fisherbrand™ 14-827-104
Beschreibung
Puncture-resistant with wide rectangular opening for horizontal loading of needles
- General-purpose sharps container with clear locking lid and marked with biohazard label
- Made from puncture-resistant polypropylene that can be autoclaved or incinerated
- Easily mount on walls and carts, nest to conserve storage space
Compliance
Used to comply with OSHA standards for sharps and biohazard waste collection.
Spezifikation
| 26.5 L | |
| Lockable | |
| 7 gal. | |
| Yes | |
| 31 cm | |
| Rectangle | |
| Autoclavable | |
| 12 in. |
| Used to comply with OSHA standards for sharps and biohazard waste collection. | |
| Clear | |
| 12 x 15.5 x 14 in. (31 x 39 x 36 cm) | |
| 36 cm | |
| 10/Cs. | |
| 39 cm | |
| 15.5 in. | |
| 14 in. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.