Learn More
Ibidi Freezing Medium Classic
Ibidi Freezing Medium Classic is a cell freezing medium for standard cell lines.
Marke: Ibidi 80023
Weitere Details : CAS : 67-68-5 Netto-Gewicht : 0.00020kg
Beschreibung
- A cell freezing medium for standard cell lines
- with Bovine Serum Albumin
- 5 x 20 mL or 120 mL
Spezifikation
Freezing Medium | |
Bovine Serum Albumin (BSA) | |
1 | |
120 mL |