Learn More
Fruquintinib, MedChemExpress
Artikelnummer. 16440428
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Packungsgröße:
5 Milligramm
50 Milligramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16440428 5 mg 5 Milligramm
16417756 100 mg -
16413897 50 mg 50 Milligramm
16407968 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16440428 Lieferant Medchem Express Lieferanten-Nr. HY199125mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Fruquintinib (HMPL-013) is a highly potent and selective VEGFR 1/2/3 inhibitor with IC50s of 33, 0.35, and 35 nM, respectively.
Fruquintinib (HMPL-013) is a highly potent and selective VEGFR 1/2/3 inhibitor with IC50s of 33, 0.35, and 35 nM, respectively.
Spezifikation
Chemischer Name oder Material Fruquintinib
CAS 1194506-26-7
Farbe White
Hinweise zur Reinheitsqualität Research
Summenformel C21H19N3O5
Menge 5 mg
Synonym HMPL-013
Löslichkeitsinformationen DMSO : 5.88 mg/mL (14.95 mM; Need ultrasonic)
SMILES O=C(C1=C(C)OC2=CC(OC3=C4C=C(OC)C(OC)=CC4=NC=N3)=CC=C12)NC
Molekulargewicht (g/mol) 393.39
Formelmasse 393.39
Reinheit (%) 99.09%
Güte Research
Physikalische Form Solid
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease