missing translation for 'onlineSavingsMsg'
Learn More

Fulleren-Pulver, hydroxyliert, C{60}(OH)n,, Thermo Scientific Chemicals

Catalog Number p-7051436
Click to view available options
Menge:
25 mg
100 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
99685-96-8
C60
720.66
MFCD06202029
XMWRBQBLMFGWIX-UHFFFAOYSA-N
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
123591
CHEBI:33128
(C60-Ih)[5,6]fullerene
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
99685-96-8
C60
720.66
MFCD06202029
XMWRBQBLMFGWIX-UHFFFAOYSA-N
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
123591
CHEBI:33128
(C60-Ih)[5,6]fullerene
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Fullerene powder
Namenshinweis Hydroxylated, C60(OH)n
Menge 25 mg
UN-Nummer UN1325
Physikalische Form Pulver

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'