missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEFT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17028-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).Spezifikation
GEFT | |
Polyclonal | |
Unconjugated | |
ARHGEF25 | |
ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115557, 314904, 52666 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P720, Q86VW2, Q9CWR0 | |
Arhgef25 | |
GEFT Fusion Protein Ag10436 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |