missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFRA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55038-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody is specific to GFRA2.
GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.Spezifikation
GFRA2 | |
Polyclonal | |
Unconjugated | |
GFRA2 | |
Gfra2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00451, O08842 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14586, 2675 | |
-20°C | |
Liquid |