Learn More
Tocris Bioscience™ GSK 650394
Serum- and glucocorticoid-regulated kinase (SGK) inhibitor
Marke: Tocris Bioscience™ 3572/50
Weitere Details : Netto-Gewicht : 0.00970kg
Beschreibung
GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).Spezifikation
>97% | |
GSK 650394 | |
GSK 650394 |
50 mg | |
382.45 |
Nur für Forschungszwecke