missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16216-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody, raised against the full-length human HBB, recognizes the 13-16 kDa HBB in western blot analyses of human heart and placenta.
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5′-epsilon - gamma-G - gamma-A - delta - beta-3′.Spezifikation
HBB | |
Polyclonal | |
Unconjugated | |
HBB | |
beta globin, CD113t C, HBB, Hemoglobin beta chain, Hemoglobin subunit beta, hemoglobin, beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15127, 3043 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P68871 | |
Hbb | |
HBB Fusion Protein Ag9216 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |