Learn More
Biomex™ Hepatitis C virus (HCV) Seroconversion Ag Only Panels
Biomex™ Hepatitis C virus (HCV) Seroconversion Ag Only Panels are intended for standard testing by diagnostic manufacturers and researchers during assay development, evaluation, troubleshooting and post-marked surveillance of HCV test systems and methods.
Marke: Biomex™ SCP-HCV-003 (Ag only)
Weitere Details : Netto-Gewicht : 0.01000kg Transport : UN number : 3373 Chem class : 6.2
Beschreibung
- Illustrates the onset and decline of immune system response to HCV from one individual over a period of time.
- Serves as validation tool for diagnostic sensitivity, determination of analytical sensitivity, identification of cut-off values or to study the humoral immune response to this infection.
- Wash hands thoroughly after handling.
- Do not eat, drink or smoke when using this product.
- Avoid the release to the environment.
Spezifikation
Single donor | |
5 x 1.0 mL Vials | |
Hepatitis C virus Ag only |