Learn More
Biomex™ HIV Seroconversion Panels
Biomex™ HIV Seroconversion Panels can be used for various research and investigation applications.
Marke: Biomex™ SCP-HIV1-005
Weitere Details : Netto-Gewicht : 0.01000kg Transport : UN number : 3373 Chem class : 6.2
Beschreibung
- Developed to aid diagnostic assay manufacturers and researchers during assay development, evaluation, troubleshooting and post-market surveillance of HIV test systems and methods.
- Can be used for validation of diagnostic assay specificity.
- For Research Use Only.
- Depicts the onset of immune system response to HIV from one individual over a period of time.
- Available defibrinated and filtered.
- Available with Sodium azide (0.095%) added as preservative.
Spezifikation
Single donor | |
25 x 1.0 mL Vials | |
−20°C to -80°C | |
HIV1 |