Learn More
Abnova™ Human A2ML1 Full-length ORF (AAH93840.1, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00144568-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The alpha-macroglobulin (AM) superfamily of proteins contains both complement components and protease inhibitors, including A2M (MIM 103950) and A2ML1. AM proteins display a unique trap mechanism of inhibition, by which the AM inhibitor undergoes a major conformational change upon its cleavage by a protease, thus trapping the protease and blocking it from subsequent substrate binding (Galliano et al., 2006 [PubMed 16298998]).[supplied by OMIM]
Sequence: MYTLEASGQGCVYVQTVLRYNILPPTNMKTFSLSVEIGKARCEQPTSPRSLTLTIHTSYVGSRSSSNMAIVEVKMLSGFSPMEGTNQLLLQQPLVKKVEFGTDTLNIYLDELIKNTQTYTFTISQSVLVTNLKPATIKVYDYYLPDEQATIQYSDPCESpezifikation
AAH93840.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
A2ML1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
144568 | |
A2ML1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPAMD9/DKFZp686C1729/DKFZp686D2011/DKFZp686G1812/DKFZp686L1821/DKFZp686O1010/FLJ16045/FLJ25179/FLJ39129/FLJ41597/FLJ41598/FLJ41607 | |
A2ML1 | |
Recombinant | |
wheat germ expression system |