Learn More
ACROBiosystems Human AIMP1 / EMAP2 / SCYE1 Protein, His Tag
Marke:  ACROBiosystems EM2-H5142-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15886637
  • 2052.84 € / 1 Milligramm
Verfügbar ab: 05-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Aminoacyl tRNA synthase complex-interacting multifunctional protein 1 (AIMP1) is also known as multisynthase complex auxiliary component p43 and endothelial monocyte-activating polypeptide II (EMAP-II). AIMP1 is a cytokine that may be induced by apoptosis and is also released from professional antigen-presenting cells such as dendritic cells. The release of AIMP1 renders the tumor-associated vasculature sensitive to tumor necrosis factor. Furthermore, AIMP1 binds tRNA and stimulates the catalytic activity of cytoplasmic arginyl-tRNA synthase. This protein possesses inflammatory cytokine activity. Also, AIMP1 negatively regulates TGF-beta signaling through stabilization of SMURF2 by binding to SMURF2 and inhibiting its SMAD7-mediated degradation.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 35.2 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
AAH14051.1
35.2 kDa
1 mg
Ala 2 - Lys 312
AIMP1,EMAP2,EMAPII,HLD3,SCYE1,p43,EMAP-2,EMAP-II,AIMP-1
Unconjugated
PBS, pH7.4
E.coli
Human
Powder
>95% as determined by SDS-PAGE
Human AIMP1, His Tag (EM2-H5142) is expressed from E.coli cells. It contains AA Ala 2 - Lys 312 (Accession # Q12904-1).
-20°C
AIMP1
His Tag
Less than 1.0 EU / µg by the LAL method.
Cytokines and Growth Factors