Learn More
ACROBiosystems Human CD209 / DC-SIGN Protein, His Tag
Marke: ACROBiosystems CD9-H5246-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD209 is also known as CLEC4L, DC-SIGN and CD209 antigen. Pathogen-recognition receptor expressed on the surface of immature dendritic cells (DCs) and involved in initiation of primary immune response. On DCs it is a high affinity receptor for ICAM2 and ICAM3 by binding to mannose-like carbohydrates. May act as a DC rolling receptor that mediates transendothelial migration of DC presursors from blood to tissues by binding endothelial ICAM2. Seems to regulate DC-induced T-cell proliferation by binding to ICAM3 on T-cells in the immunological synapse formed between DC and T-cells.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 41.3 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_066978.1 | |
1 mg | |
Gln 59 - Ala 404 | |
DC-SIGN,CD209,DC-SIGN1,CLEC4L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
41.3 kDa | |
Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession # Q9NN x 6-1). | |
-20°C | |
CD209 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |