missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD209 / DC-SIGN Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_066978.1 |
---|---|
Molekulargewicht | 41.3 kDa |
Quelle | Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession # Q9NN x 6-1). |
Immunogen | Gln 59 - Ala 404 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16457339
|
ACROBiosystems
CD9-H5246-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 27-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16447339
|
ACROBiosystems
CD9-H5246-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 27-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
CD209 is also known as CLEC4L, DC-SIGN and CD209 antigen. Pathogen-recognition receptor expressed on the surface of immature dendritic cells (DCs) and involved in initiation of primary immune response. On DCs it is a high affinity receptor for ICAM2 and ICAM3 by binding to mannose-like carbohydrates. May act as a DC rolling receptor that mediates transendothelial migration of DC presursors from blood to tissues by binding endothelial ICAM2. Seems to regulate DC-induced T-cell proliferation by binding to ICAM3 on T-cells in the immunological synapse formed between DC and T-cells.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 41.3 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_066978.1 | |
Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession # Q9NN x 6-1). | |
-20°C | |
CD209 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
41.3 kDa | |
Gln 59 - Ala 404 | |
DC-SIGN,CD209,DC-SIGN1,CLEC4L | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |