Learn More
ACROBiosystems Human CD300c / LMIR2 Protein, His Tag
Marke: ACROBiosystems CDC-H5224-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
CMRF35-like molecule 6 (CLM-6) is also known as CD300 antigen-like family member C, CMRF35-A1, Immunoglobulin superfamily member 16 (IgSF16) , CD antigen CD300c, which belongs to the CD300 family. CLM-6 contains one Ig-like V-type (immunoglobulin-like) domain. CD300a/c RNA and surface expression were downregulated after stimulation of plasmacytoid dendritic cells (pDCs) with TLR7 and TLR9 ligands. Exogenous interferon (IFN)-alpha down-regulated CD300a/c expression. CD300a and CD300c play an important role in the cross-regulation of TNF-alpha and IFN-alpha secretion from pDCs. CD300a and CD300c are indistinguishable on the surface of NK cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.7 kDa. The protein migrates as 26-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH22279 | |
100 μg | |
Gly 21 - Arg 183 | |
CD300C,CLM-6,CMRF35-A1,CMRF-35,CMRF35A,IGSF16 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
18.7 kDa | |
Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Arg 183 (Accession # AAH22279). | |
-20°C | |
CD300c | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |