Learn More
ACROBiosystems Human CD69 / CLEC2C Protein, His Tag
Marke:  ACROBiosystems CD9-H5222-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15896317
  • 2007.22 € / 1 Milligramm
Verfügbar ab: 05-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Early activation antigen CD69 is also known as C-type lectin domain family 2 member C (CLEC2C), Activation inducer molecule (AIM), Early T-cell activation antigen p60, Leukocyte surface antigen Leu-23. CD69 contains one C-type lectin domain. CD69 / CLEC2C is expressed on the surface of activated T-cells, B-cells, natural killer cells, neutrophils, eosinophils, epidermal Langerhans cells and platelets. CD69 is induced by antigens, mitogens or activators of PKC on the surface of T and B-lymphocytes and by interaction of IL-2 with the p75 IL-2R on the surface of NK cells. CD69 / CLEC2C involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes, natural killer (NK) cells, and platelets.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20-26 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Spezifikation
Spezifikation
AAH07037
16.8 kDa
1 mg
Ser 62 - Lys 199
CD69,CLEC2C,AIM,BL-AC/P26,EA1,GP32/28,MLR-3
Unconjugated
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). It contains AA Ser 62 - Lys 199 (Accession # AAH07037).
-20°C
CD69
His Tag
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens