missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD94 Protein, His Tag

Marke:  ACROBiosystems CD4-H52H8-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16497289

  • 483.87 € / 100 Mikrogramm
Verfügbar ab: 27-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD94(Natural killer cells antigen CD94) is also known as KLRD1, KP43, NK cell receptor, Killer cell lectin-like receptor subfamily D member 1. CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.1 kDa. The protein migrates as 24-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

19.1 kDa
Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1).
-20°C
CD94
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens
100 μg
Lys 32 - Ile 179
CD94,KLRD1,KP43,NK cell receptor
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers