missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD94 Protein, His Tag
483.87 € - 3495.90 €
Spezifikation
Molekulargewicht | 19.1 kDa |
---|---|
Quelle | Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1). |
Immunogen | Lys 32 - Ile 179 |
Lagerungsbedingungen | -20°C |
Gen-Alias | CD94,KLRD1,KP43,NK cell receptor |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16497289
|
ACROBiosystems
CD4-H52H8-100UG |
100 μg |
483.87 €
100 Mikrogramm |
Verfügbar ab: 08-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16407299
|
ACROBiosystems
CD4-H52H8-1MG |
1 mg |
3495.90 €
1 Milligramm |
Verfügbar ab: 08-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
CD94(Natural killer cells antigen CD94) is also known as KLRD1, KP43, NK cell receptor, Killer cell lectin-like receptor subfamily D member 1. CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.1 kDa. The protein migrates as 24-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
19.1 kDa | |
Lys 32 - Ile 179 | |
CD94,KLRD1,KP43,NK cell receptor | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Ile 179 (Accession # Q13241-1). | |
-20°C | |
CD94 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |