Learn More
Abnova™ Human CENPH Full-length ORF (AAH12024, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00064946-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Centromere and kinetochore proteins play a critical role in centromere structure, kinetochore formation, and sister chromatid separation. The protein encoded by this gene colocalizes with inner kinetochore plate proteins CENP-A and CENP-C in both interphase and metaphase. It localizes outside of centromeric heterochromatin, where CENP-B is localized, and inside the kinetochore corona, where CENP-E is localized during prometaphase. It is thought that this protein can bind to itself, as well as to CENP-A, CENP-B or CENP-C. Multimers of the protein localize constitutively to the inner kinetochore plate and play an important role in the organization and function of the active centromere-kinetochore complex. [provided by RefSeq]
Sequence: MEEQPQMQDADEPADSGGEGRAGGPPQVAGAQAACSEDRMTLLLRLRAQTKQQLLEYKSMVDASEEKTPEQIMQEKQIEAKIEDLENEIEEVKVAFEIKKLALDRMRLSTALKKNLEKISRQSSVLMDNMKHLLELNKLIMKSQQESWDLEEKLLDIRKKRLQLKQASESKLLEIQTEKNKQKIDLDSMENSERIKIIRQNLQMEIKITTVIQHVFQNLILGSKVNWAEDPALKEIVLQLEKNVDMMSpezifikation
AAH12024 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.91kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NNF1/PMF1 | |
CENPH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64946 | |
CENPH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEEQPQMQDADEPADSGGEGRAGGPPQVAGAQAACSEDRMTLLLRLRAQTKQQLLEYKSMVDASEEKTPEQIMQEKQIEAKIEDLENEIEEVKVAFEIKKLALDRMRLSTALKKNLEKISRQSSVLMDNMKHLLELNKLIMKSQQESWDLEEKLLDIRKKRLQLKQASESKLLEIQTEKNKQKIDLDSMENSERIKIIRQNLQMEIKITTVIQHVFQNLILGSKVNWAEDPALKEIVLQLEKNVDMM | |
RUO | |
CENPH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |