Learn More
Abnova™ Human CREBL2 Partial ORF (NP_001301, 31 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00001389-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
cAMP response element (CRE)-binding protein-like-2 (CREBL2) was identified in a search to find genes in a commonly deleted region on chromosome 12p13 flanked by ETV6 and CDKN1B genes, frequently associated with hematopoietic malignancies, as well as breast, non-small-cell lung and ovarian cancers. CREBL2 shares a 41% identity with CRE-binding protein (CREB) over a 48-base long region which encodes the bZip domain of CREB. The bZip domain consists of about 30 amino acids rich in basic residues involved in DNA binding, followed by a leucine zipper motif involved in protein dimerization. This suggests that CREBL2 encodes a protein with DNA binding capabilities. The occurance of CREBL2 deletion in malignancy suggests that CREBL2 may act as a tumor suppressor gene. [provided by RefSeq]
Sequence: ERSRQSARECRARKKLRYQYLEELVSSRERAICALREELEMYKQWCMAMDQGKIPSEIKALLTGEEQNKSQQNSSRHTKAGKTDANSNSWSpezifikation
NP_001301 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ERSRQSARECRARKKLRYQYLEELVSSRERAICALREELEMYKQWCMAMDQGKIPSEIKALLTGEEQNKSQQNSSRHTKAGKTDANSNSW | |
RUO | |
CREBL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1389 | |
CREBL2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC117311/MGC138362 | |
CREBL2 | |
Recombinant | |
wheat germ expression system |