Learn More
ACROBiosystems Human Dkk-3 (R335G) Protein, His Tag
Marke: ACROBiosystems DK3-H5220-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Members of the dickkopf-related protein family (DKK-1, -2, -3, and -4) are secreted proteins with two cysteine-rich domains separated by a linker region. And DKK3 has been proposed as tumour suppressor gene and a marker for tumour blood vessels. DKK3 is the only DKK family member abundantly expressed in normal lung, but silenced by promoter hypermethylation in a large fraction of lung cancer cell lines and lung tumors. Downregulation of DKK3 was correlated with tumor progression and expression of nuclear beta-catenin in lung tumors. Ectopic expression of DKK3 in lung cancer cells with DKK3 hypermethylation induced apoptosis and inhibited TCF-4 activity as well as nuclear accumulation of beta-catenin and expression of TCF-4 targets c-Myc and cyclin D1. DKK3 modulates FGF and Activin/Nodal signaling to regulate mesoderm induction during early Xenopus development, was reported.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.0 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001018067.1 (R335G) | |
200 μg | |
Ala 22 - Ile 350 | |
DKK3,REIC,RIG | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
37.0 kDa | |
Human Dkk-3 (R335G), His Tag (DK3-H5220) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Ile 350 (Accession # Q9UBP4-1 (R335G)). | |
-20°C | |
Dkk-3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |