Learn More
ACROBiosystems Human DR6 / TNFRSF21 Protein, Fc Tag
Marke: ACROBiosystems TN1-H5252-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 21 (TNFRSF21) is also known as death receptor 6 (DR6), which is a member of the TNF-receptor superfamily. TNFRSF21 contains one death domain and four TNFR-Cys repeats. TNFRSF21 / DR6 has been shown to activate NF-κB and MAPK8/JNK, and induce cell apoptosis. Through its death domain, this receptor interacts with TRADD protein, which is known to serve as an adaptor that mediates signal transduction of TNF-receptors.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 80-95kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH17730 | |
1 mg | |
Gln 42 - Leu 350 | |
TNFRSF21,DR6,CD358,BM-018 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
60.2 kDa | |
Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession # AAH17730). | |
-20°C | |
DR6 | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |