Learn More
Abnova™ Human GDI2 Full-length ORF (AAH05145.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002665-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MNEEYDVIVLGTGLTECILSGIMSVNGKKVLHMDRNPYYGGESASITPLEDLYKRFKIPGSPPESMERGRDWNVDLIPKFLMANGQLVKMLLYTEVTRYLDFKVTEGSFVYKGGKIYKVPSTEAEALASSLMGLFEKRRFRKFLVYVANFDEKDPRTFEGIDPKKTTMRDVYKKFDLGQDVIDFTGHALALYRTDDYLDQPCYETINRIKLYSESLARYGKSPYLYPLYGLGELPQGFARLGAIYGGTYMLNKPIEEIIVQNGKVIGVKSEGEIARCKQLICDPSYVKDRVEKVGQVIRVICILSHPIKNTNDANSCQIIIPQNQVNRKSDIYVCMISFAHNVAAQGKYIAIVSTTVETKEPEKEIRPALELLEPIEQKFVSISDLLVPKDLGTESQIFISRTYDATTHFETTCDDIKNIYKRMTGSEFDFEEMKRKKNDIYGEDSpezifikation
AAH05145.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.69kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16452/FLJ37352/RABGDIB | |
GDI2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2665 | |
GDI2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNEEYDVIVLGTGLTECILSGIMSVNGKKVLHMDRNPYYGGESASITPLEDLYKRFKIPGSPPESMERGRDWNVDLIPKFLMANGQLVKMLLYTEVTRYLDFKVTEGSFVYKGGKIYKVPSTEAEALASSLMGLFEKRRFRKFLVYVANFDEKDPRTFEGIDPKKTTMRDVYKKFDLGQDVIDFTGHALALYRTDDYLDQPCYETINRIKLYSESLARYGKSPYLYPLYGLGELPQGFARLGAIYGGTYMLNKPIEEIIVQNGKVIGVKSEGEIARCKQLICDPSYVKDRVEKVGQVIRVICILSHPIKNTNDANSCQIIIPQNQVNRKSDIYVCMISFAHNVAAQGKYIAIVSTTVETKEPEKEIRPALELLEPIEQKFVSISDLLVPKDLGTESQIFISRTYDATTHFETTCDDIKNIYKRMTGSEFDFEEMKRKKNDIYGED | |
RUO | |
GDI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |