missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GDI2 Full-length ORF (AAH05145.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00002665-P01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16185411

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 24-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

GDP dissociation inhibitors are proteins that regulate the GDP-GTP exchange reaction of members of the rab family, small GTP-binding proteins of the ras superfamily, that are involved in vesicular trafficking of molecules between cellular organelles. GDIs slow the rate of dissociation of GDP from rab proteins and release GDP from membrane-bound rabs. GDI2 is ubiquitously expressed. The GDI2 gene contains many repetitive elements indicating that it may be prone to inversion/deletion rearrangements. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MNEEYDVIVLGTGLTECILSGIMSVNGKKVLHMDRNPYYGGESASITPLEDLYKRFKIPGSPPESMERGRDWNVDLIPKFLMANGQLVKMLLYTEVTRYLDFKVTEGSFVYKGGKIYKVPSTEAEALASSLMGLFEKRRFRKFLVYVANFDEKDPRTFEGIDPKKTTMRDVYKKFDLGQDVIDFTGHALALYRTDDYLDQPCYETINRIKLYSESLARYGKSPYLYPLYGLGELPQGFARLGAIYGGTYMLNKPIEEIIVQNGKVIGVKSEGEIARCKQLICDPSYVKDRVEKVGQVIRVICILSHPIKNTNDANSCQIIIPQNQVNRKSDIYVCMISFAHNVAAQGKYIAIVSTTVETKEPEKEIRPALELLEPIEQKFVSISDLLVPKDLGTESQIFISRTYDATTHFETTCDDIKNIYKRMTGSEFDFEEMKRKKNDIYGED
Spezifikation

Spezifikation

AAH05145.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.69kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16452/FLJ37352/RABGDIB
GDI2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2665
GDI2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNEEYDVIVLGTGLTECILSGIMSVNGKKVLHMDRNPYYGGESASITPLEDLYKRFKIPGSPPESMERGRDWNVDLIPKFLMANGQLVKMLLYTEVTRYLDFKVTEGSFVYKGGKIYKVPSTEAEALASSLMGLFEKRRFRKFLVYVANFDEKDPRTFEGIDPKKTTMRDVYKKFDLGQDVIDFTGHALALYRTDDYLDQPCYETINRIKLYSESLARYGKSPYLYPLYGLGELPQGFARLGAIYGGTYMLNKPIEEIIVQNGKVIGVKSEGEIARCKQLICDPSYVKDRVEKVGQVIRVICILSHPIKNTNDANSCQIIIPQNQVNRKSDIYVCMISFAHNVAAQGKYIAIVSTTVETKEPEKEIRPALELLEPIEQKFVSISDLLVPKDLGTESQIFISRTYDATTHFETTCDDIKNIYKRMTGSEFDFEEMKRKKNDIYGED
RUO
GDI2
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote