Learn More
ACROBiosystems Human IFN-gamma R1 / IFNGR1 Protein, Fc Tag
Marke: ACROBiosystems IF1-H5254-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interferon gamma receptor 1 (IFNGR1) is also known as CD antigen CD119, which belongs to the type II cytokine receptor family. IFNGR1 contains two fibronectin type-III domains and two Ig-like C2-type (immunoglobulin-like) domains. IFNGR1 / CD119 is receptor for interferon gamma. Two receptors bind one interferon gamma (IFNG) dimer.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.4 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH05333 | |
1 mg | |
Glu 18 - Gly 245 | |
IFNGR1,CD119,CDw119,IFNGR,IFN-gamma-R1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
52.4 kDa | |
Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333). | |
-20°C | |
IFN-gamma R1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |