Learn More
Abnova™ Human IMPG2 Partial ORF (NP_057331.2, 572 a.a. - 678 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00050939-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interphotoreceptor matrix proteoglycan-2 is part of an extracellular complex occupying the interface between photoreceptors and the retinal pigment epithelium in the fundus of the eye.[supplied by OMIM]
Sequence: LTSKVKDQLKVSPFLPDASMEKELIFDGGLGSGSGQKVDLITWPWSETSSEKSAEPLSKPWLEDDDSLLPAEIEDKKLVLVDKMDSTDQISKHSKYEHDDRSTHFPESpezifikation
NP_057331.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LTSKVKDQLKVSPFLPDASMEKELIFDGGLGSGSGQKVDLITWPWSETSSEKSAEPLSKPWLEDDDSLLPAEIEDKKLVLVDKMDSTDQISKHSKYEHDDRSTHFPE | |
RUO | |
IMPG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50939 | |
IMPG2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IPM200/SPACRCAN | |
IMPG2 | |
Recombinant | |
wheat germ expression system |