Learn More
ACROBiosystems Human M-CSF R / CSF1R / CD115 Protein, Mouse IgG2a Fc Tag
MALS verified
Marke: ACROBiosystems CSR-H5255-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Colony stimulating factor 1 receptor (CSF1R) is also known as macrophage colony-stimulating factor receptor (M-CSFR), CD115 Cluster of Differentiation 115 (CD115), C-FMS, CSFR, FIM2, FMS, and is a member of the typeIII subfamily of receptor tyrosine kinases (RTKs). CSF1R is a receptor for a cytokine called colony stimulating factor 1, The protein encoded by the CSFR1 gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most, if not all, of the biological effects of this cytokine. Ligand binding activates CSFR1 through a process of oligomerization and transphosphorylation . Mutations in CSF1R are associated with chronic myelomonocytic leukemia and type M4 acute myeloblastic leukemia. Increased levels of CSF1R1 are found in microglia in Alzheimer's disease and after brain injuries. The increased receptor expression causes microglia to become more active. Both CSF1R, and its ligand colony stimulating factor 1 play an important role in the development of the mammary gland and may be involved in the process of mammary gland carcinogenesis.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 81.4 kDa. The protein migrates as 105-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_005202.2 | |
100 μg | |
Ile 20 - Glu 512 | |
CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
81.4 kDa | |
Human M-CSF R, Mouse IgG2a Fc Tag (MALS verified) (CSR-H5255) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # P07333-1). | |
-20°C | |
M-CSF R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |