missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MIR16 Partial ORF (NP_057725, 31 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00051573-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: ACLLTGSLFVLLRVFSFEPVPSCRALQVLKPRDRISAIAHRGGSHDAPENTLAAIRQAAKNGATGVELDIEFTSDGIPVLMHDNTVDRTTDGTGRLCDLTFEQIRKLNSpezifikation
NP_057725 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ACLLTGSLFVLLRVFSFEPVPSCRALQVLKPRDRISAIAHRGGSHDAPENTLAAIRQAAKNGATGVELDIEFTSDGIPVLMHDNTVDRTTDGTGRLCDLTFEQIRKLN | |
RUO | |
GDE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51573 | |
MIR16 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
363E6.2/MIR16 | |
GDE1 | |
Recombinant | |
wheat germ expression system |