missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NR2F2 Partial ORF (NP_066285, 153 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007026-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: RMPPTQPTHGQFALTNGDPLNCHSYLSGYISLLLRAEPYPTSRFGSQCMQPNNIMGIENICELAARMLFSAVEWARNIPFFPDLQITDSpezifikation
NP_066285 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RMPPTQPTHGQFALTNGDPLNCHSYLSGYISLLLRAEPYPTSRFGSQCMQPNNIMGIENICELAARMLFSAVEWARNIPFFPDLQITD | |
RUO | |
NR2F2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7026 | |
NR2F2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARP1/COUP-TFII/COUPTFB/MGC117452/SVP40/TFCOUP2 | |
NR2F2 | |
Recombinant | |
wheat germ expression system |