Learn More
Abnova™ Human PCGF6 Full-length ORF (AAH07602.1, 1 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084108-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene contains a RING finger motif, which is most closely related to those of polycomb group (PcG) proteins RNF110/MEL-18 and BMI1. PcG proteins are known to form protein complexes and function as transcription repressors. This protein has been shown to interact with some PcG proteins and act as a transcription repressor. The activity of this protein is found to be regulated by cell cycle dependent phosphorylation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MEGVAVVTAGSVGAAKTEGAAALPPPPPPPVSPPALTPAPAAGEEGPAPLSETGAPGCSGSRPPELEPERSLGRFRGRFEDEDEELEEEEELEEEEEEEEEDMSHFSLRLEGGRQDSEDEEERLINLSELTPYILCSICKGYLIDATTITECLHTFCKSCIVRHFYYSNRCPKCNIVVHQTQPLYNISANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQVDIICGDHLLEQYQTLREIRRAIGDAAMQDGLLVLHYGLVVSPLKITSpezifikation
AAH07602.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MBLR/MGC15678/MGC17541/RNF134 | |
PCGF6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84108 | |
PCGF6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGVAVVTAGSVGAAKTEGAAALPPPPPPPVSPPALTPAPAAGEEGPAPLSETGAPGCSGSRPPELEPERSLGRFRGRFEDEDEELEEEEELEEEEEEEEEDMSHFSLRLEGGRQDSEDEEERLINLSELTPYILCSICKGYLIDATTITECLHTFCKSCIVRHFYYSNRCPKCNIVVHQTQPLYNISANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQVDIICGDHLLEQYQTLREIRRAIGDAAMQDGLLVLHYGLVVSPLKIT | |
RUO | |
PCGF6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |