Learn More
ACROBiosystems Human PSMA / FOLH1 Protein, Fc Tag
Marke: ACROBiosystems PSA-H5264-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Prostate-specific membrane antigen (PSMA) is also known as Folate hydrolase 1 (FOLH1), Glutamate carboxypeptidase 2 (GCP2), N-acetylated-alpha-linked acidic dipeptidase I (NAALAD1), which belongs to the peptidase M28 family and M28B subfamily. FOLH1 / PSMA is stable at pH greater than 6.5. FOLH1 / PSMA is a type II transmembrane zinc metallopeptidase that is most highly expressed in the nervous system, prostate, kidney, and small intestine. FOLH1 / GCP-2 is homodimer and binds 2 zinc ions per subunit, and required for NAALADase activity. The catalytic activity of PSMA involved in releasing of an unsubstituted, C-terminal glutamyl residue, typically from Ac-Asp-Glu or folylpoly – gamma - glutamates. FOLH1 / GCP-2 / PSMA has both folate hydrolase and N – acetylated – alpha – linked - acidic dipeptidase (NAALADase) activity and has a preference for tri-alpha-glutamate peptides. GCP-2 / PSMA involved in prostate tumor progression and also exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 106.0 kDa. The protein migrates as 110-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_058050.3 | |
100 μg | |
Lys 44 - Ala 750 | |
FOLH1,PSMA,GIG27,FOLH,NAALAD1,PSM,NAALADase I,GCPII,FGCP | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
106.0 kDa | |
Human PSMA, Fc Tag (PSA-H5264) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1). | |
-20°C | |
PSMA | |
Human | |
50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |