Learn More
STEMCELL Technologies Human Recombinant Activin A
Activin A
Marke:  STEMCELL Technologies 78001
Weitere Details : Netto-Gewicht : 0.00110kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 17169311
  • 240.30 € / 10 Mikrogramm
Verfügbar ab: 12-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
  • Activin A is a member of the transforming growth factor beta (TGF-β) family of proteins produced by many cell types throughout development (Gurdon et al.) It is a disulfide-linked homodimer (two beta-A chains) that binds to heteromeric complexes of a type I (Act RI-A and Act RI-B) and a type II (Act RII-A and Act RII-B) serine-threonine kinase receptor (Attisano et al.) Activins primarily signal through SMAD2/3 proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (McDowell et al.) Activin A maintains the undifferentiated state of human embryonic stem cells (James et al.; Xiao et al.) and also facilitates differentiation of human embryonic stem cells into definitive endoderm (D'Amour et al.)
Spezifikation
Spezifikation
Activin beta-A chain, EDF, Erythroid differentiation protein, FRP, FSH-releasing protein, INHBA, Inhibin beta-A chain, Inhibin beta-1
10 μg
Endoderm, PSC-Derived; Mesoderm, PSC-Derived; Other; Pluripotent Stem Cells
≥95%