Learn More
Abnova™ Human RTN2 Partial ORF (NP_005610, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006253-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MGQVLPVFAHCKEAPSTASSTPDSTEGGNDDSDFRELHTAREFSEEDEEETTSQDWGTPRELTFSYIAFDGVVGSGGRRDSTARRPRPQGRSVSEPRDQHSpezifikation
NP_005610 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGQVLPVFAHCKEAPSTASSTPDSTEGGNDDSDFRELHTAREFSEEDEEETTSQDWGTPRELTFSYIAFDGVVGSGGRRDSTARRPRPQGRSVSEPRDQH | |
RUO | |
RTN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6253 | |
RTN2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NSP2/NSPL1 | |
RTN2 | |
Recombinant | |
wheat germ expression system |