missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Serpin D1 / Heparin Cofactor II Protein, His Tag
350.80 € - 2419.31 €
Spezifikation
Zugriffsnummer | AAH35028 |
---|---|
Molekulargewicht | 55.7 kDa |
Quelle | Human Serpin D1, His Tag (HC2-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Ser 499 (Accession # AAH35028). |
Immunogen | Gly 20 - Ser 499 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15806927
|
ACROBiosystems
HC2-H5224-1MG |
1 mg |
2419.31 €
1 Milligramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15816927
|
ACROBiosystems
HC2-H5224-50UG |
50 μg |
350.80 €
50 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Serpin D1 is also known as Heparin cofactor 2 (HCF2), Protease inhibitor leuserpin-2 (HLS2), Heparin cofactor II (HC-II), which belongs to the serpin family. HCF2 is expressed predominantly in liver. Thrombin inhibitor activated by the glycosaminoglycans, heparin or dermatan sulfate. In the presence of the latter, SerpinD1 / HC-II becomes the predominant thrombin inhibitor in place of antithrombin III (AT-III). In a glycosaminoglycan-independent manner, serpinD1 / HC-II also inhibits chymotrypsin. Peptides at the N-terminal of HC-II have chemotactic activity for both monocytes and neutrophils.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.7 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH35028 | |
Human Serpin D1, His Tag (HC2-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Ser 499 (Accession # AAH35028). | |
-20°C | |
Serpin D1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH 7.5 | |
HEK293 | |
Other Recombinant Proteins |
55.7 kDa | |
Gly 20 - Ser 499 | |
SerpinD1,HCF2,HLS2,HC-II | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |