Learn More
ACROBiosystems Human Serpin E2 / PN1 Protein, His Tag
Marke: ACROBiosystems SE2-H5225-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Description
SERPINE2 is also known as Glia-derived nexin (GDN), Peptidase inhibitor 7 (PI7), Protease nexin 1(PN1). SERPINE2 is a secreted glycoprotein which belongs to the serpin family. SerpinE1 is the primary physiological inhibitor of the two plasminogen activators urokinase (uPA) and tissue plasminogen activator (tPA). PAI-1 / GDN is also implicated in adipose tissue development. It suggests that PAI-1 inhibitors serve in the control of atherothrombosis. Defects in Serpin E1 / PN1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency) which is characterized by abnormal bleeding due to SerpinE1 defect in the plasma.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.8 kDa. The protein migrates as 48-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
42.8 kDa | |
Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Pro 397 (Accession # P07093-2). | |
-20°C | |
Serpin E2 | |
Human | |
20 mM Sodium Acetate, 100 mM NaCl, pH6.5 | |
HEK293 | |
Other Recombinant Proteins |
25 μg | |
Ser 20 - Pro 397 | |
SERPINE2,Glia-derived nexin,GDN,Peptidase inhibitor 7,PI-7,Protease nexin 1,PN-1,Serpin E2,SerpinE2,PI7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |