missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Serpin E2 / PN1 Protein, His Tag
350.80 € - 1959.64 €
Spezifikation
Molekulargewicht | 42.8 kDa |
---|---|
Quelle | Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Pro 397 (Accession # P07093-2). |
Immunogen | Ser 20 - Pro 397 |
Lagerungsbedingungen | -20°C |
Gen-Alias | SERPINE2,Glia-derived nexin,GDN,Peptidase inhibitor 7,PI-7,Protease nexin 1,PN-1,Serpin E2,SerpinE2,PI7 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15897667
|
ACROBiosystems
SE2-H5225-500UG |
500 μg |
1959.64 €
500 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15887667
|
ACROBiosystems
SE2-H5225-25UG |
25 μg |
350.80 €
25 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
SERPINE2 is also known as Glia-derived nexin (GDN), Peptidase inhibitor 7 (PI7), Protease nexin 1(PN1). SERPINE2 is a secreted glycoprotein which belongs to the serpin family. SerpinE1 is the primary physiological inhibitor of the two plasminogen activators urokinase (uPA) and tissue plasminogen activator (tPA). PAI-1 / GDN is also implicated in adipose tissue development. It suggests that PAI-1 inhibitors serve in the control of atherothrombosis. Defects in Serpin E1 / PN1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency) which is characterized by abnormal bleeding due to SerpinE1 defect in the plasma.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.8 kDa. The protein migrates as 48-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
42.8 kDa | |
Ser 20 - Pro 397 | |
SERPINE2,Glia-derived nexin,GDN,Peptidase inhibitor 7,PI-7,Protease nexin 1,PN-1,Serpin E2,SerpinE2,PI7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Human Serpin E2, His Tag (SE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Pro 397 (Accession # P07093-2). | |
-20°C | |
Serpin E2 | |
Human | |
20 mM Sodium Acetate, 100 mM NaCl, pH6.5 | |
HEK293 | |
Other Recombinant Proteins |