missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SNX14 Partial ORF (AAH05110, 784 a.a. - 893 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00057231-Q01.25ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16172613

  • 508.00 € / 25 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. The encoded protein also contains a regulator of G protein signaling (RGS) domain. Regulator of G protein signaling family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWM
Spezifikation

Spezifikation

AAH05110
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QLFQEHRLVSLITLLRDAIFCENTEPRSLQDKQKGAKQTFEEMMNYIPDLLVKCIGEETKYESIRLLFDGLQQPVLNKQLTYVLLDIVIQELFPELNKVQKEVTSVTSWM
RUO
SNX14
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57231
SNX14 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC13217/RGS-PX2
SNX14
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote