missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human TROP-2 / TACSTD2 Protein, His Tag
350.80 € - 1512.06 €
Spezifikation
Zugriffsnummer | NP_002344.2 |
---|---|
Molekulargewicht | 29.8 kDa |
Quelle | Human TROP-2, His Tag (TR2-H5223) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1). |
Immunogen | His 27 - Thr 274 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15837857
|
ACROBiosystems
TR2-H5223-1MG |
1 mg |
1512.06 €
1 Milligramm |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15847857
|
ACROBiosystems
TR2-H5223-200UG |
200 μg |
350.80 €
200 Mikrogramm |
Verfügbar ab: 23-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
TROP-2 is a single-copy gene in human cells, and encodes a type-1 transmembrane glycoprotein which is over-expressed in various malignancies, also referred to as tumor associated calcium signal transducer 2 (TACSTD2), GA733-1 or M1S1. TROP-2 is related to epithelial cell adhesion molecule (EpCAM), also called TROP-1, gp40, and KSA. Trop-1 and Trop-2 are homologous to serum IGF-II-binding proteins and appear as signal transducers. Thus, they likely represent novel cell-surface receptors and may play a role in regulating the growth of carcinoma cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.8 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002344.2 | |
Human TROP-2, His Tag (TR2-H5223) is expressed from human 293 cells (HEK293). It contains AA His 27 - Thr 274 (Accession # P09758-1). | |
-20°C | |
TROP-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
29.8 kDa | |
His 27 - Thr 274 | |
TACSTD2,GA733-1,M1S1,TROP2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |