Learn More
Abnova™ Human UPK1B Partial ORF (NP_008883, 131 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007348-Q01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is found in the asymmetrical unit membrane (AUM) where it can form a complex with other transmembrane 4 superfamily proteins. It may play a role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq]
Sequence: NNSPPNNDDQWKNNGVTKTWDRLMLQDNCCGVNGPSDWQKYTSAFRTENNDADYPWPRQCCVMNNLKEPLNLEACKLGVPGFYHNQGCYELISGPMNRSpezifikation
NP_008883 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NNSPPNNDDQWKNNGVTKTWDRLMLQDNCCGVNGPSDWQKYTSAFRTENNDADYPWPRQCCVMNNLKEPLNLEACKLGVPGFYHNQGCYELISGPMNR | |
RUO | |
UPK1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7348 | |
UPK1B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TSPAN20/UPIB/UPK1 | |
UPK1B | |
Recombinant | |
wheat germ expression system |