missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO5L, MedChemExpress
Description
IDO5L is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 67 nM.
Specifications
Specifications
| Chemischer Name oder Material | IDO5L |
| CAS | 914471-09-3 |
| Farbe | Light Yellow |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C9H7ClFN5O2 |
| Menge | 500 mg |
| Löslichkeitsinformationen | DMSO : ≥ 52 mg/mL (191.43 mM) |
| SMILES | ON/C(C1=NON=C1N)=N/C2=CC=C(F)C(Cl)=C2 |
| Molekulargewicht (g/mol) | 271.64 |
| Formelmasse | 271.64 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?