missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-8 (1-77) (CXCL8) Monoclonal Antibody (8CH), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience 53-8088-41
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse Monoclonal Antibody
IL-8 (1-77) (CXCL8) Monoclonal antibody specifically detects IL-8 (1-77) (CXCL8) in Human samples. It is validated for Flow CytometrySpezifikation
IL-8 (1-77) (CXCL8) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P10145 | |
CXCL8 | |
Affinity chromatography | |
RUO | |
3576 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
8CH | |
Alexa Fluor 488 | |
CXCL8 | |
AMCF-I; ANX7; BOS_6776; Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; CXCL8; Emoctakin; GCP1; GCP-1; Granulocyte chemotactic protein 1; IL8; IL-8; ILN; Interleukin; Interleukin8; Interleukin-8; LECT; LUCT; LYNAP; MDNCF; MONAP; Monocyte-derived neutrophil chemotactic factor; Monocyte-derived neutrophil-activating peptide; NAF; NAP1; NAP-1; Neutrophil attractant/activation protein 1; Neutrophil-activating protein 1; Permeability factor 1; PF1; Protein 3-10C; RP11-537A6.8; SNX; SYNEXIN; T-cell chemotactic factor | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |