missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-8 (1-77) (CXCL8) Monoclonal Antibody (8CH), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
112.00 € - 273.00 €
Spezifikation
Antigen | IL-8 (1-77) (CXCL8) |
---|---|
Klon | 8CH |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16341004
|
Affymetrix eBioscience
53-8088-42 |
100 Tests |
273.00 €
100 Stück |
Verfügbar ab: 05-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16331004
|
Affymetrix eBioscience
53-8088-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Mouse Monoclonal Antibody
Interleukin 8 (IL-8, CXCL8) is a 72 amino acid pro-inflammatory factor which belongs to the CXC subfamily of chemokines, and are bound by the cell surface receptors IL-8RA and IL-8RB. IL-8 functions as a chemoattractant and potent angiogenic factor. The expression and secretion of IL-8 can be induced by diverse inflammatory stimuli in many cells, including macrophages and endothelial cells. In endothelial cells, IL-8 is present in storage vesicles called Weibel-Palade bodies. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C. IL-8 and ten other members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours, and IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection.Spezifikation
IL-8 (1-77) (CXCL8) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
3576 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
8CH | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
P10145 | |
CXCL8 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CXCL8 |