missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IRAK4, His Tag Recombinant Protein
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases.
Marke: Invitrogen™ PV3362
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
IL-1 receptor associated kinases (IRAKs) are serine⁄threonine kinases involved in signaling via Toll⁄IL-1 pathways.Spezifikation
NP_057207 | |
51135 | |
10 μg | |
IRAK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
Z'-LYTE™ | |
interleukin-1 receptor-associated kinase 4 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
55.6 kDa | |
IRAK4 | |
Human | |
His Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
IRAK4 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |