missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Monoclonal Antibody (2F1), Super Bright™ 600, eBioscience™, Invitrogen™
Syrian Hamster Monoclonal Antibody
Marke: Affymetrix eBioscience 63-5893-80
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Syrian Hamster Monoclonal Antibody
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow CytometrySpezifikation
KLRG1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
O88713 | |
Klrg1 | |
Affinity chromatography | |
RUO | |
50928 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2F1 | |
Super Bright 600 | |
Klrg1 | |
2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; ITIM-containing receptor MAFA-L; Killer cell lectin-like receptor subfamily G member 1; Klrg1; MAFA; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; Mast cell function-associated antigen 2F1 | |
Syrian Hamster | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |