Learn More
DWK Life Sciences Kimble™ Kontes™ Screw Thread Culture Tubes with Unattached Rubber Lined Caps
Uniform wall thickness for maximum heat transfer and heat resistance
472.00 €
Beschreibung
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- KG-33 borosilicate glass
- Designed from ASTM™ E982, Type VI, Class A requirements
- Thread finish complies with Glass Packaging Institute (GPI) requirements
- Cap is not attached